missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycinnamic acid, 99%

CAS: 830-09-1 | C10H10O3 | 178.187 g/mol

$122.16 - $335.41

Chemical Identifiers

CAS 830-09-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00004398
InChI Key AFDXODALSZRGIH-QPJJXVBHSA-N
Synonym 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
PubChem CID 699414
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(C=C1)C=CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6014414
View Documents
Thermo Scientific Chemicals
H6014414
25 g
Each for $122.16
Only null left
 
AAH6014422
View Documents
Thermo Scientific Chemicals
H6014422
100 g
Each for $335.41
Only null left
 
Description

Description

It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool dry conditions. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

830-09-1
178.187
AFDXODALSZRGIH-QPJJXVBHSA-N
699414
COC1=CC=C(C=C1)C=CC(=O)O
C10H10O3
MFCD00004398
4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
(E)-3-(4-methoxyphenyl)prop-2-enoic acid

Specifications

830-09-1
C10H10O3
25 g
Soluble in dimethyl sulfoxide and methanol.
COC1=CC=C(C=C1)C=CC(=O)O
178.187
178.18
4-Methoxycinnamic acid, predominantly trans
170°C to 173°C
MFCD00004398
4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
AFDXODALSZRGIH-QPJJXVBHSA-N
(E)-3-(4-methoxyphenyl)prop-2-enoic acid
699414
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-594-0

RTECSNumber : UD3391300

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only