missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-alpha-methylstyrene (stabilized with TBC) 98.0+%, TCI America™
$160.30 - $464.14
Chemical Identifiers
CAS | 350-40-3 |
---|---|
Molecular Formula | C9H9F |
Molecular Weight (g/mol) | 136.169 |
MDL Number | MFCD00042297 |
InChI Key | VIXHMBLBLJSGIB-UHFFFAOYSA-N |
Synonym | 4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene |
PubChem CID | 67690 |
IUPAC Name | 1-fluoro-4-prop-1-en-2-ylbenzene |
SMILES | CC(=C)C1=CC=C(C=C1)F |
Chemical Identifiers
350-40-3 | |
136.169 | |
VIXHMBLBLJSGIB-UHFFFAOYSA-N | |
67690 | |
CC(=C)C1=CC=C(C=C1)F |
C9H9F | |
MFCD00042297 | |
4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene | |
1-fluoro-4-prop-1-en-2-ylbenzene |
Specifications
350-40-3 | |
C9H9F | |
5 mL | |
4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene | |
CC(=C)C1=CC=C(C=C1)F | |
136.169 | |
136.17 | |
Liquid |
183°C | |
MFCD00042297 | |
1993 | |
VIXHMBLBLJSGIB-UHFFFAOYSA-N | |
1-fluoro-4-prop-1-en-2-ylbenzene | |
67690 | |
≥98.0% (GC) | |
4-Fluoro-alpha-methylstyrene (stabilized with TBC) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator