missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylaminocinnamaldehyde 98.0+%, TCI America™
$130.48 - $378.40
Chemical Identifiers
CAS | 6203-18-5 |
---|---|
Molecular Formula | C11H13NO |
Molecular Weight (g/mol) | 175.231 |
MDL Number | MFCD00007002 |
InChI Key | RUKJCCIJLIMGEP-ONEGZZNKSA-N |
Synonym | 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent |
PubChem CID | 5284506 |
IUPAC Name | (E)-3-[4-(dimethylamino)phenyl]prop-2-enal |
SMILES | CN(C)C1=CC=C(C=C1)C=CC=O |
Chemical Identifiers
6203-18-5 | |
175.231 | |
RUKJCCIJLIMGEP-ONEGZZNKSA-N | |
5284506 | |
CN(C)C1=CC=C(C=C1)C=CC=O |
C11H13NO | |
MFCD00007002 | |
4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent | |
(E)-3-[4-(dimethylamino)phenyl]prop-2-enal |
Specifications
6203-18-5 | |
C11H13NO | |
5 g | |
RUKJCCIJLIMGEP-ONEGZZNKSA-N | |
(E)-3-[4-(dimethylamino)phenyl]prop-2-enal | |
5284506 | |
≥98.0% (T) | |
4-Dimethylaminocinnamaldehyde |
140°C | |
MFCD00007002 | |
4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent | |
CN(C)C1=CC=C(C=C1)C=CC=O | |
175.231 | |
175.23 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1187
TSCA : Yes