Learn More
4-Chloropyridine hydrochloride, 98+%
CAS: 7379-35-3 | C5H5Cl2N | 150.00 g/mol
Supplier: Thermo Scientific Chemicals A1562814
Description
It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
4-Chloropyridine hydrochloride | |
∼210°C (sublimation) | |
198°C (388°F) | |
C5H5Cl2N | |
25 g | |
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride | |
XGAFCCUNHIMIRV-UHFFFAOYSA-N | |
4-chloropyridine;hydrochloride | |
81852 | |
≥98% |
7379-35-3 | |
2 | |
Weak | |
MFCD00012829 | |
3684904 | |
Soluble in water. | |
[H+].[Cl-].ClC1=CC=NC=C1 | |
150.00 | |
150.01 |
Chemical Identifiers
7379-35-3 | |
150.00 | |
XGAFCCUNHIMIRV-UHFFFAOYSA-N | |
81852 | |
[H+].[Cl-].ClC1=CC=NC=C1 |
C5H5Cl2N | |
MFCD00012829 | |
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride | |
4-chloropyridine;hydrochloride |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 230-946-1
RTECSNumber : US6300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only