missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Biphenylacetic Acid 98.0+%, TCI America™
$76.43 - $432.46
Chemical Identifiers
CAS | 5728-52-9 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.248 |
MDL Number | MFCD00004351 |
InChI Key | QRZAKQDHEVVFRX-UHFFFAOYSA-N |
Synonym | 4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid |
PubChem CID | 3332 |
ChEBI | CHEBI:31597 |
IUPAC Name | 2-(4-phenylphenyl)acetic acid |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O |
Chemical Identifiers
5728-52-9 | |
212.248 | |
QRZAKQDHEVVFRX-UHFFFAOYSA-N | |
3332 | |
2-(4-phenylphenyl)acetic acid |
C14H12O2 | |
MFCD00004351 | |
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid | |
CHEBI:31597 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O |
Specifications
5728-52-9 | |
White-Yellow | |
MFCD00004351 | |
2811 | |
QRZAKQDHEVVFRX-UHFFFAOYSA-N | |
2-(4-phenylphenyl)acetic acid | |
3332 | |
212.25 | |
Crystalline Powder |
163°C | |
C14H12O2 | |
25 g | |
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid | |
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O | |
212.248 | |
CHEBI:31597 | |
≥98.0% (HPLC,T) | |
4-Biphenylacetic Acid |
Safety and Handling
RTECSNumber : DU8229050
TSCA : No