missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenyl Benzoate 98.0+%, TCI America™

$262.83 - $898.48

Chemical Identifiers

CAS 2170-13-0
Molecular Formula C19H14O2
Molecular Weight (g/mol) 274.319
MDL Number MFCD00223541
InChI Key CINHWMYRCOGYIX-UHFFFAOYSA-N
Synonym Benzoic Acid 4-Biphenyl Ester
PubChem CID 231743
IUPAC Name (4-phenylphenyl) benzoate
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)OC(=O)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18665G
View Documents
TCI America
B18665G
5 g
Each for $262.83
Only null left
 
B186625G
View Documents
TCI America
B186625G
25 g
Each for $898.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2170-13-0
274.319
CINHWMYRCOGYIX-UHFFFAOYSA-N
231743
C1=CC=C(C=C1)C2=CC=C(C=C2)OC(=O)C3=CC=CC=C3
C19H14O2
MFCD00223541
Benzoic Acid 4-Biphenyl Ester
(4-phenylphenyl) benzoate

Specifications

2170-13-0
White
MFCD00223541
Benzoic Acid 4-Biphenyl Ester
C1=CC=C(C=C1)C2=CC=C(C=C2)OC(=O)C3=CC=CC=C3
274.319
274.32
Crystalline Powder
151°C
C19H14O2
5 g
CINHWMYRCOGYIX-UHFFFAOYSA-N
(4-phenylphenyl) benzoate
231743
≥98.0% (GC)
4-Biphenyl Benzoate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'