Learn More
4-Benzyloxy-3-methoxybenzaldehyde, 98%
CAS: 2426-87-1 | C15H14O3 | 242.27 g/mol
$101.11 - $280.97
Chemical Identifiers
CAS | 2426-87-1 |
---|---|
Molecular Formula | C15H14O3 |
Molecular Weight (g/mol) | 242.27 |
MDL Number | MFCD00003365 |
InChI Key | JSHLOPGSDZTEGQ-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde |
PubChem CID | 75506 |
IUPAC Name | 3-methoxy-4-phenylmethoxybenzaldehyde |
SMILES | COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1471614
|
Thermo Scientific Chemicals
A1471614 |
25 g |
N/A
|
|
|||||
AAA1471622
|
Thermo Scientific Chemicals
A1471622 |
100 g |
N/A
|
|
|||||
Description
4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N?-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N′-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.
Solubility
Soluble in chloroform, methanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong bases.
Chemical Identifiers
2426-87-1 | |
242.27 | |
JSHLOPGSDZTEGQ-UHFFFAOYSA-N | |
75506 | |
COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1 |
C15H14O3 | |
MFCD00003365 | |
4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde | |
3-methoxy-4-phenylmethoxybenzaldehyde |
Specifications
2426-87-1 | |
213°C to 215°C (3.5 mmHg) | |
MFCD00003365 | |
1464258 | |
4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde | |
JSHLOPGSDZTEGQ-UHFFFAOYSA-N | |
3-methoxy-4-phenylmethoxybenzaldehyde | |
75506 | |
98% |
61°C to 65°C | |
C15H14O3 | |
25 g | |
Air Sensitive | |
Soluble in chloroform,methanol. | |
COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1 | |
242.27 | |
242.27 | |
4-Benzyloxy-3-methoxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 219-379-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only