missing translation for 'onlineSavingsMsg'
Learn More

4,6-O-(4-Methoxybenzylidene)-D-glucal, 97%, Thermo Scientific™

$73.26 - $206.53

Chemical Identifiers

CAS 312623-79-3
Molecular Formula C14H16O5
Molecular Weight (g/mol) 264.277
MDL Number MFCD01863528
InChI Key SSKZQXMNNREVNP-HABKJSAYSA-N
Synonym 4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
PubChem CID 46185760
IUPAC Name (4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60633MD
View Documents
Thermo Scientific Chemicals
H60633MD
250 mg
Each for $73.26
Only null left
 
AAH6063303
View Documents
Thermo Scientific Chemicals
H6063303
1 g
Each for $206.53
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,6-O-(4-Methoxybenzylidene)-D-glucal is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Chemical Identifiers

312623-79-3
264.277
SSKZQXMNNREVNP-HABKJSAYSA-N
46185760
COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O
C14H16O5
MFCD01863528
4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
(4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol

Specifications

312623-79-3
MFCD01863528
4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O
264.277
264.27
4,6-O-(4-Methoxybenzylidene)-D-glucal
C14H16O5
250 mg
SSKZQXMNNREVNP-HABKJSAYSA-N
(4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
46185760
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'