missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic acid, 98%

CAS: 4998-07-6 | C9H8NO6 | 226.17 g/mol

Supplier:  Thermo Scientific Chemicals L0851014

 View more versions of this product

Catalog No. AAL0851014


Only null left
Explore available promotions

Description

Description

4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.

Solubility
Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

4,5-Dimethoxy-2-nitrobenzoic acid
192°C to 195°C
MFCD00014697
2123903
Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).
C1CCN(C1)C2CCNCC2
154.26
227.17
4998-07-6
C9H18N2
25 g
4-1-pyrrolidinyl piperidine, 4-pyrrolidin-1-yl piperidine, 4-pyrrolidin-1-yl-piperidine, 4-pyrrolidinopiperidine, 4-1-pyrrolidinyl-piperidine, piperidine, 4-1-pyrrolidinyl, 4-pyrrolidinylpiperidine, 4-pyrrolizinopiperidine, 4-pyrrolidino-piperidine
STWODXDTKGTVCJ-UHFFFAOYSA-N
4-pyrrolidin-1-ylpiperidine
78703
98%

Chemical Identifiers

4998-07-6
MFCD00014697
4-pyrrolidin-1-ylpiperidine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-657-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only