missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4',5'-Dibromofluorescein, Thermo Scientific™
$36.55 - $36.55
Chemical Identifiers
CAS | 596-03-2 |
---|---|
Molecular Formula | C20H10Br2O5 |
Molecular Weight (g/mol) | 490.11 |
InChI Key | ZDTNHRWWURISAA-UHFFFAOYSA-N |
Synonym | 4',5'-dibromofluorescein, solvent red 72, eosinic acid, japan orange 201, FD&C orange no. 5, c.i. solvent red 72, 4',5'-dibromofluororescein, 4,5-dibromofluorescein, d & c orange no. 5, japan orange no. 201 |
PubChem CID | 11689 |
IUPAC Name | 4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |
SMILES | C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC202430050
|
Thermo Scientific Chemicals
202430050 |
5 g |
Each for $36.55
|
|
|||||
Chemical Identifiers
596-03-2 | |
490.11 | |
4',5'-dibromofluorescein, solvent red 72, eosinic acid, japan orange 201, FD&C orange no. 5, c.i. solvent red 72, 4',5'-dibromofluororescein, 4,5-dibromofluorescein, d & c orange no. 5, japan orange no. 201 | |
4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |
C20H10Br2O5 | |
ZDTNHRWWURISAA-UHFFFAOYSA-N | |
11689 | |
C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O |
Specifications
596-03-2 | |
5% max. | |
Glass bottle | |
5 g | |
15,3027 | |
(1% in hot 0.5 N NH4OH) Clear yellow to orange | |
C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O | |
490.11 | |
490.11 |
270.0°C to 273.0°C | |
Authentic | |
C20H10Br2O5 | |
19,V,6,462 | |
4',5'-dibromofluorescein, solvent red 72, eosinic acid, japan orange 201, FD&C orange no. 5, c.i. solvent red 72, 4',5'-dibromofluororescein, 4,5-dibromofluorescein, d & c orange no. 5, japan orange no. 201 | |
ZDTNHRWWURISAA-UHFFFAOYSA-N | |
4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one | |
11689 | |
4',5'-Dibromofluorescein |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 209-876-
RUO – Research Use Only