missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxychalcone, 99%

CAS: 2373-89-9 | C17H16O3 | 268.31 g/mol

$98.68 - $333.64

Chemical Identifiers

CAS 2373-89-9
Molecular Formula C17H16O3
Molecular Weight (g/mol) 268.31
MDL Number MFCD00025815
InChI Key HDXVSZWKIHQDES-LFYBBSHMSA-N
Synonym 4,4'-dimethoxychalcone, e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 2e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 1,3-bis 4-methoxyphenyl prop-2-en-1-one, 4,4/'-dimethoxychalcone, 2-propen-1-one, 1,3-bis 4-methoxyphenyl, e-4,4'-dimethoxychalcone, 1,3-bis 4-methoxyphenyl-2-propen-1-one, e-1,3-bis 4-methoxyphenyl-2-propen-1-one, 2e-1,3-bis 4-methoxyphenyl-2-propen-1-one
PubChem CID 5377817
IUPAC Name (E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1058509
View Documents
Thermo Scientific Chemicals
L1058509
10 g
Each for $98.68
Only null left
 
AAL1058518
View Documents
Thermo Scientific Chemicals
L1058518
50 g
Each for $333.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2373-89-9
268.31
HDXVSZWKIHQDES-LFYBBSHMSA-N
5377817
COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1
C17H16O3
MFCD00025815
4,4'-dimethoxychalcone, e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 2e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 1,3-bis 4-methoxyphenyl prop-2-en-1-one, 4,4/'-dimethoxychalcone, 2-propen-1-one, 1,3-bis 4-methoxyphenyl, e-4,4'-dimethoxychalcone, 1,3-bis 4-methoxyphenyl-2-propen-1-one, e-1,3-bis 4-methoxyphenyl-2-propen-1-one, 2e-1,3-bis 4-methoxyphenyl-2-propen-1-one
(E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one

Specifications

2373-89-9
C17H16O3
10 g
4,4'-dimethoxychalcone, e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 2e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 1,3-bis 4-methoxyphenyl prop-2-en-1-one, 4,4/'-dimethoxychalcone, 2-propen-1-one, 1,3-bis 4-methoxyphenyl, e-4,4'-dimethoxychalcone, 1,3-bis 4-methoxyphenyl-2-propen-1-one, e-1,3-bis 4-methoxyphenyl-2-propen-1-one, 2e-1,3-bis 4-methoxyphenyl-2-propen-1-one
COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1
268.31
268.32
4,4'-Dimethoxychalcone
99°C to 103°C
MFCD00025815
1622293
HDXVSZWKIHQDES-LFYBBSHMSA-N
(E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
5377817
99%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only