missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorocyclohexanecarboxylic Acid 98.0+%, TCI America™

$57.79 - $188.27

Chemical Identifiers

CAS 122665-97-8
Molecular Formula C7H10F2O2
Molecular Weight (g/mol) 164.152
MDL Number MFCD03788493
InChI Key HYIUDFLDFSIXTR-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
PubChem CID 2779136
IUPAC Name 4,4-difluorocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D43301G
View Documents
TCI America
D43301G
1 g
Each for $57.79
Only null left
 
D43305G
View Documents
TCI America
D43305G
5 g
Each for $188.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

122665-97-8
164.152
HYIUDFLDFSIXTR-UHFFFAOYSA-N
2779136
C1CC(CCC1C(=O)O)(F)F
C7H10F2O2
MFCD03788493
4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
4,4-difluorocyclohexane-1-carboxylic acid

Specifications

122665-97-8
White
MFCD03788493
4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
C1CC(CCC1C(=O)O)(F)F
164.152
164.15
Crystalline Powder
107°C
C7H10F2O2
1 g
HYIUDFLDFSIXTR-UHFFFAOYSA-N
4,4-difluorocyclohexane-1-carboxylic acid
2779136
≥98.0% (GC,T)
4,4-Difluorocyclohexanecarboxylic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'