Learn More
3M™ E-A-R™ Yellow Neons™ Metal Detectable Ear Plugs
E-A-R™ metal detectable ear plugs have a stainless steel bead encased in the ear plug. Popular in food manufacturing industries, when contamination prevention is critical.
Supplier:  3M™ ACA3114106
Catalog No. 29-635-1074
Only null left
Explore available promotions
Description
Description
  • Designed with detectable metal components for food processing and other industries
  • Low-pressure in the ear canal for enhanced comfort
  • Poly bag keeps each pair of ear plugs clean before use
  • Moisture-resistant helps maintain a consistent expansion time, even in humid situations
  • Slightly longer than regular size 3M™ E-A-R™ Classic™ ear plugs for higher noise reduction
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Class AL
Food Processing, Manufacturing, Pulp and Paper, Pharmaceutical, Tobacco Processing Industries and Other Various Industries That Require Detectable Products to Prevent Contamination
311-4106
2000 Pairs/Cs
Yellow
Corded
Single-use
Blue
Regular
Yes
Poly bag
Foam
32 dB
Bullet
Yes
Product Suggestions
Product Suggestions