Learn More
Corning™ 3D Clear Penetration Buffer
Corning™'s 3D Clear Penetration Buffer is used with the Corning™ 3D immunolabeling process and is comprised of PBS/0.2% Triton™ X-100/0.3 M glycine/20% DMSO.
$463.17 - $463.17
Specifications
Includes | PBS/0.2% Triton™ X-100/0.3 M glycine/20% DMSO |
---|---|
Content And Storage | For best results, refrigerate after opening |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10-320-112
|
Corning™
5739 |
100 mL |
Case for $463.17
|
|
|||||
Description
- Rapid tissue clearing
- Easy to Use
- No Special Equipment Required
- Compatible with IF, FP and other Fluorescent labels
- Microplate and Automation Compatible
- Reversible to Follow up with 2D H&E/IHC
Specifications
PBS/0.2% Triton™ X-100/0.3 M glycine/20% DMSO | |
For best results, refrigerate after opening |