missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Sulfolene 98.0+%, TCI America™
$42.87 - $108.11
Chemical Identifiers
CAS | 77-79-2 |
---|---|
Molecular Formula | C4H6O2S |
Molecular Weight (g/mol) | 118.15 |
MDL Number | MFCD00005481 |
InChI Key | MBDNRNMVTZADMQ-UHFFFAOYSA-N |
Synonym | 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569 |
PubChem CID | 6498 |
IUPAC Name | 2,5-dihydrothiophene 1,1-dioxide |
SMILES | C1C=CCS1(=O)=O |
Chemical Identifiers
77-79-2 | |
118.15 | |
MBDNRNMVTZADMQ-UHFFFAOYSA-N | |
6498 | |
C1C=CCS1(=O)=O |
C4H6O2S | |
MFCD00005481 | |
3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569 | |
2,5-dihydrothiophene 1,1-dioxide |
Specifications
77-79-2 | |
White-Yellow | |
MFCD00005481 | |
3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569 | |
C1C=CCS1(=O)=O | |
118.15 | |
118.15 | |
Crystalline Powder |
65°C | |
C4H6O2S | |
25 g | |
MBDNRNMVTZADMQ-UHFFFAOYSA-N | |
2,5-dihydrothiophene 1,1-dioxide | |
6498 | |
≥98.0% (GC) | |
3-Sulfolene |
Safety and Handling
EINECSNumber : (5)-5264
RTECSNumber : XM9100000
TSCA : Yes