missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinonehydrazone Hydrochloride Hydrate 98.0+%, TCI America™

$34.02 - $85.03

Chemical Identifiers

CAS 149022-15-1
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.70
MDL Number MFCD00149370
InChI Key OEZPVSPULCMUQB-UHFFFAOYSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
PubChem CID 9575839
IUPAC Name 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
SMILES Cl.CN1C(SC2=CC=CC=C12)=NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21551G
View Documents
TCI America
M21551G
1 g
Each for $34.02
Only null left
 
M21555G
View Documents
TCI America
M21555G
5 g
Each for $85.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

149022-15-1
215.70
OEZPVSPULCMUQB-UHFFFAOYSA-N
9575839
Cl.CN1C(SC2=CC=CC=C12)=NN
C8H10ClN3S
MFCD00149370
unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride

Specifications

149022-15-1
MFCD00149370
2811
OEZPVSPULCMUQB-UHFFFAOYSA-N
2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
9575839
≥98.0% (HPLC,T)
3-Methyl-2-benzothiazolinonehydrazone Hydrochloride Hydrate [for Biochemical Research]
C8H10ClN3S
1 g
unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
Cl.CN1C(SC2=CC=CC=C12)=NN
215.70
215.70
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DL7160200

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'