Learn More
3-Methoxy-o-phenylenediamine, 97%
CAS: 37466-89-0 | C7H10N2O | 138.17 g/mol
$337.18 - $430.74
Chemical Identifiers
CAS | 37466-89-0 |
---|---|
Molecular Formula | C7H10N2O |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD08276903 |
InChI Key | BFLWXPJTAKXXKT-UHFFFAOYSA-N |
Synonym | 2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline |
PubChem CID | 184268 |
IUPAC Name | 3-methoxybenzene-1,2-diamine |
SMILES | COC1=CC=CC(=C1N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH35286MD
|
Thermo Scientific Chemicals
H35286MD |
250 mg |
Each for $337.18
|
|
|||||
AAH3528603
|
Thermo Scientific Chemicals
H3528603 |
1 g |
Each for $430.74
|
|
|||||
Description
3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
Chemical Identifiers
37466-89-0 | |
138.17 | |
BFLWXPJTAKXXKT-UHFFFAOYSA-N | |
184268 | |
COC1=CC=CC(=C1N)N |
C7H10N2O | |
MFCD08276903 | |
2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline | |
3-methoxybenzene-1,2-diamine |
Specifications
37466-89-0 | |
C7H10N2O | |
250 mg | |
Air sensitive | |
Slightly soluble in water. | |
COC1=CC=CC(=C1N)N | |
138.17 | |
138.17 | |
3-Methoxy-o-phenylenediamine |
39°C to 40°C | |
MFCD08276903 | |
UN2811 | |
2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline | |
BFLWXPJTAKXXKT-UHFFFAOYSA-N | |
3-methoxybenzene-1,2-diamine | |
184268 | |
97% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only