Learn More
3-Hydroxy-4-methoxybenzoic acid, 97%
CAS: 645-08-9 | C8H8O4 | 168.148 g/mol
Supplier: Thermo Scientific Chemicals A1370914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Hydroxy-4-methoxybenzoic acid | |
251°C to 255°C | |
MFCD00002507 | |
2208365 | |
LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
3-hydroxy-4-methoxybenzoic acid | |
12575 | |
168.15 |
645-08-9 | |
C8H8O4 | |
25 g | |
isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
COC1=C(C=C(C=C1)C(=O)O)O | |
168.148 | |
CHEBI:63798 | |
≥97% |
Chemical Identifiers
645-08-9 | |
168.148 | |
LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
12575 | |
3-hydroxy-4-methoxybenzoic acid |
C8H8O4 | |
MFCD00002507 | |
isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
CHEBI:63798 | |
COC1=C(C=C(C=C1)C(=O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-430-5
RTECSNumber : BZ4850000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only