missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(pentafluorothio)aniline, 97%

CAS: 1240256-99-8 | C6H5F6NS | 237.163 g/mol

$331.88 - $1082.14

Chemical Identifiers

CAS 1240256-99-8
Molecular Formula C6H5F6NS
Molecular Weight (g/mol) 237.163
MDL Number MFCD16652430
InChI Key RFXWDLYDFAZNLO-UHFFFAOYSA-N
Synonym 3-fluoro-5-pentafluorosulfur aniline, 3-fluoro-5-pentafluoro-??-sulfanyl aniline, 3-fluoro-5-pentafluorothio aniline
PubChem CID 66523473
IUPAC Name 3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)aniline
SMILES C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33680MD
View Documents
Thermo Scientific Chemicals
H33680MD
250 mg
Each for $331.88
Only null left
 
AAH3368003
View Documents
Thermo Scientific Chemicals
H3368003
1 g
Each for $1,082.14
Only null left
 
Description

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1240256-99-8
237.163
RFXWDLYDFAZNLO-UHFFFAOYSA-N
66523473
C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)N
C6H5F6NS
MFCD16652430
3-fluoro-5-pentafluorosulfur aniline, 3-fluoro-5-pentafluoro-??-sulfanyl aniline, 3-fluoro-5-pentafluorothio aniline
3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)aniline

Specifications

1240256-99-8
MFCD16652430
UN2811
Sparingly soluble in water.(0.26g/L) (25°C),
C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)N
237.163
237.16
3-Fluoro-5-(pentafluorothio)aniline
C6H5F6NS
250 mg
3-fluoro-5-pentafluorosulfur aniline, 3-fluoro-5-pentafluoro-??-sulfanyl aniline, 3-fluoro-5-pentafluorothio aniline
RFXWDLYDFAZNLO-UHFFFAOYSA-N
3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)aniline
66523473
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only