missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothieno[3,2-b]thiophene 94.0+%, TCI America™

$473.48 - $1394.32

Chemical Identifiers

CAS 25121-83-9
Molecular Formula C6H3BrS2
Molecular Weight (g/mol) 219.114
InChI Key YFWCQBIKGWTZQQ-UHFFFAOYSA-N
PubChem CID 14383679
IUPAC Name 6-bromothieno[3,2-b]thiophene
SMILES C1=CSC2=C1SC=C2Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B3226200MG
View Documents
TCI America
B3226200MG
200 mg
Each for $473.48
Only null left
 
B32261G
View Documents
TCI America
B32261G
1 g
Each for $1,394.32
Only null left
 
Description

Description

Specifications

Chemical Identifiers

25121-83-9
219.114
14383679
C1=CSC2=C1SC=C2Br
C6H3BrS2
YFWCQBIKGWTZQQ-UHFFFAOYSA-N
6-bromothieno[3,2-b]thiophene

Specifications

25121-83-9
C6H3BrS2
YFWCQBIKGWTZQQ-UHFFFAOYSA-N
6-bromothieno[3,2-b]thiophene
14383679
≥94.0% (GC)
3-Bromothieno[3,2-b]thiophene
Yellow
200 mg
C1=CSC2=C1SC=C2Br
219.114
219.11
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'