missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopropyltrimethoxysilane 96.0+%, TCI America™
$41.02 - $251.65
Chemical Identifiers
CAS | 13822-56-5 |
---|---|
Molecular Formula | C6H17NO3Si |
Molecular Weight (g/mol) | 179.291 |
MDL Number | MFCD00008206 |
InChI Key | SJECZPVISLOESU-UHFFFAOYSA-N |
Synonym | 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6 |
PubChem CID | 83756 |
IUPAC Name | 3-trimethoxysilylpropan-1-amine |
SMILES | CO[Si](CCCN)(OC)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T125525ML
|
TCI America
T125525ML |
25 mL |
Each for $41.02
|
|
|||||
T1255100ML
|
TCI America
T1255100ML |
100 mL |
Each for $85.75
|
|
|||||
T1255500ML
|
TCI America
T1255500ML |
500 mL |
Each for $251.65
|
|
|||||
Chemical Identifiers
13822-56-5 | |
179.291 | |
SJECZPVISLOESU-UHFFFAOYSA-N | |
83756 | |
CO[Si](CCCN)(OC)OC |
C6H17NO3Si | |
MFCD00008206 | |
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6 | |
3-trimethoxysilylpropan-1-amine |
Specifications
13822-56-5 | |
92°C | |
MFCD00008206 | |
1760 | |
SJECZPVISLOESU-UHFFFAOYSA-N | |
3-trimethoxysilylpropan-1-amine | |
83756 | |
≥96.0% (GC,T) | |
3-Aminopropyltrimethoxysilane |
Yellow | |
C6H17NO3Si | |
25 mL | |
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6 | |
CO[Si](CCCN)(OC)OC | |
179.291 | |
179.29 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-2061
TSCA : Yes