Learn More
3-Aminobenzeneboronic acid hemisulfate, 98+%
CAS: 66472-86-4 | C12H18B2N2O8S | 371.96 g/mol
Supplier: Thermo Scientific Chemicals A1724006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminobenzeneboronic acid hemisulfate | |
>300°C | |
MFCD00013111 | |
8094151 | |
3-aminophenyl boronic acid sulfate 2:1, 3-aminobenzeneboronic acid hemisulfate salt, 3-aminophenylboronic acid hemisulfate, 3-aminobenzeneboronic acid hemisulfate, 3-aminophenylboronic acid hemisulphate, 4-aminophenylboronic acid hemisulfate, m-aminophenyl boronic acid, hemisulphate, boronic acid, 3-aminophenyl-, sulfate 2:1, 3-aminophenyl boronic acid; sulfuric acid, bis m-aminophenylboronic acid ; sulfuric acid | |
OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O | |
371.96 | |
185.98 |
66472-86-4 | |
C12H18B2N2O8S | |
5 g | |
Hygroscopic | |
UKTAURVTSWDIQR-UHFFFAOYSA-N | |
(3-aminophenyl)boronic acid;sulfuric acid | |
16211139 | |
≥98% |
Chemical Identifiers
66472-86-4 | |
371.96 | |
UKTAURVTSWDIQR-UHFFFAOYSA-N | |
16211139 | |
OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O |
C12H18B2N2O8S | |
MFCD00013111 | |
3-aminophenyl boronic acid sulfate 2:1, 3-aminobenzeneboronic acid hemisulfate salt, 3-aminophenylboronic acid hemisulfate, 3-aminobenzeneboronic acid hemisulfate, 3-aminophenylboronic acid hemisulphate, 4-aminophenylboronic acid hemisulfate, m-aminophenyl boronic acid, hemisulphate, boronic acid, 3-aminophenyl-, sulfate 2:1, 3-aminophenyl boronic acid; sulfuric acid, bis m-aminophenylboronic acid ; sulfuric acid | |
(3-aminophenyl)boronic acid;sulfuric acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 266-376-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only