missing translation for 'onlineSavingsMsg'
Learn More

3,5-Heptanedione 97.0+%, TCI America™

$136.06 - $410.08

Chemical Identifiers

CAS 7424-54-6
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015186
InChI Key DGCTVLNZTFDPDJ-UHFFFAOYSA-N
Synonym 3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
PubChem CID 81923
IUPAC Name heptane-3,5-dione
SMILES CCC(=O)CC(=O)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13955G
View Documents
TCI America
H13955G
5 g
Each for $136.06
Only null left
 
H139525G
View Documents
TCI America
H139525G
25 g
Each for $410.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7424-54-6
128.171
DGCTVLNZTFDPDJ-UHFFFAOYSA-N
81923
CCC(=O)CC(=O)CC
C7H12O2
MFCD00015186
3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
heptane-3,5-dione

Specifications

7424-54-6
177°C
MFCD00015186
1224
DGCTVLNZTFDPDJ-UHFFFAOYSA-N
heptane-3,5-dione
81923
≥97.0% (GC)
3,5-Heptanedione
Yellow
C7H12O2
5 g
3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
CCC(=O)CC(=O)CC
128.171
128.17
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3141

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'