missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)aniline, 98+%

CAS: 14268-66-7 | C7H7NO2 | 137.138 g/mol

Supplier:  Thermo Scientific Chemicals A1297518

 View more versions of this product

Catalog No. AAA1297518


Only null left
Explore available promotions

Description

Description

3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong acids, acid chlorides, acid anhydrides. Air & Light Sensitive. Stable under recommended storage conditions.
Specifications

Specifications

3,4-(Methylenedioxy)aniline
40°C to 45°C
142°C (287°F)
MFCD00005832
UN2811
Air and light sensitive
Slightly soluble in water.
C1OC2=C(O1)C=C(C=C2)N
137.138
137.14
14268-66-7
144°C (16 mmHg)
C7H7NO2
50 g
4919
3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene
XGNXYCFREOZBOL-UHFFFAOYSA-N
1,3-benzodioxol-5-amine
84310
≥98%

Chemical Identifiers

14268-66-7
137.138
XGNXYCFREOZBOL-UHFFFAOYSA-N
84310
C1OC2=C(O1)C=C(C=C2)N
C7H7NO2
MFCD00005832
3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene
1,3-benzodioxol-5-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 238-161-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only