missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%

CAS: 883738-38-3 | C18H27BN2O3 | 330.24 g/mol

$188.89 - $533.12

Chemical Identifiers

CAS 883738-38-3
Molecular Formula C18H27BN2O3
Molecular Weight (g/mol) 330.24
MDL Number MFCD05864308
InChI Key ZYDMNNQZEUSDGG-UHFFFAOYSA-N
Synonym 4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
PubChem CID 2760029
SMILES CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2922403
View Documents
Thermo Scientific Chemicals
H2922403
1 g
Each for $188.89
Only null left
 
AAH2922406
View Documents
Thermo Scientific Chemicals
H2922406
5 g
Each for $533.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

883738-38-3
330.24
ZYDMNNQZEUSDGG-UHFFFAOYSA-N
2760029
C18H27BN2O3
MFCD05864308
4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

883738-38-3
C18H27BN2O3
1 g
ZYDMNNQZEUSDGG-UHFFFAOYSA-N
(4-methylpiperazin-1-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
2760029
97%
86°C to 89°C
MFCD05864308
4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
330.24
330.23
3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only