missing translation for 'onlineSavingsMsg'
Learn More

3,4-Lutidine, 98+%

CAS: 583-58-4 | C7H9N | 107.156 g/mol

Supplier:  Thermo Scientific Chemicals L03489AE

 View more versions of this product

Catalog No. AAL03489AE


Only null left
Explore available promotions

Description

Description

3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.

Solubility
Miscible with water, ethanol and ether.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
Specifications

Specifications

3,4-Lutidine
-11°C
163°C to 164°C
Unpleasant
1.512
100 mL
106583
Miscible with water,ethanol and ether.
CC1=C(C=NC=C1)C
107.156
107.16
583-58-4
0.955
47°C (116°F)
C7H9N
MFCD00006403
UN2929
3,4-lutidine, pyridine, 3,4-dimethyl, unii-fcu895kj7h, pyridine,3,4-dimethyl, fcu895kj7h, 3,4-lutidene, t 3,4-lutidine, 3,4-dimethyl-pyridine, 5-20-06-00054 beilstein handbook reference
NURQLCJSMXZBPC-UHFFFAOYSA-N
3,4-dimethylpyridine
11417
≥98%

Chemical Identifiers

583-58-4
107.156
NURQLCJSMXZBPC-UHFFFAOYSA-N
3,4-dimethylpyridine
C7H9N
MFCD00006403
11417
CC1=C(C=NC=C1)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H310-H331-H226-H302-H315-H319-H335
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H332-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

EINECSNumber : 209-511-5

RTECSNumber : OK9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only