missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene 95.0+%, TCI America™
$70.84 - $430.60
Chemical Identifiers
CAS | 84-19-5 |
---|---|
Molecular Formula | C22H22O4 |
Molecular Weight (g/mol) | 350.414 |
MDL Number | MFCD00063286 |
InChI Key | YWLLGDVBTLPARJ-LWKKFVLGSA-N |
Synonym | Dienestrol Diacetate |
PubChem CID | 24884313 |
IUPAC Name | [4-[(2Z,4E)-4-(4-acetyloxyphenyl)hexa-2,4-dien-3-yl]phenyl] acetate |
SMILES | CC=C(C1=CC=C(C=C1)OC(=O)C)C(=CC)C2=CC=C(C=C2)OC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B1366100MG
|
TCI America
B1366100MG |
100 mg |
Each for $70.84
|
|
|||||
B13661G
|
TCI America
B13661G |
1 g |
Each for $430.60
|
|
|||||
Chemical Identifiers
84-19-5 | |
350.414 | |
YWLLGDVBTLPARJ-LWKKFVLGSA-N | |
24884313 | |
CC=C(C1=CC=C(C=C1)OC(=O)C)C(=CC)C2=CC=C(C=C2)OC(=O)C |
C22H22O4 | |
MFCD00063286 | |
Dienestrol Diacetate | |
[4-[(2Z,4E)-4-(4-acetyloxyphenyl)hexa-2,4-dien-3-yl]phenyl] acetate |
Specifications
84-19-5 | |
White-Yellow | |
MFCD00063286 | |
Dienestrol Diacetate | |
CC=C(C1=CC=C(C=C1)OC(=O)C)C(=CC)C2=CC=C(C=C2)OC(=O)C | |
350.414 | |
350.41 | |
Crystalline Powder |
122°C | |
C22H22O4 | |
100 mg | |
YWLLGDVBTLPARJ-LWKKFVLGSA-N | |
[4-[(2Z,4E)-4-(4-acetyloxyphenyl)hexa-2,4-dien-3-yl]phenyl] acetate | |
24884313 | |
≥95.0% (GC) | |
3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene |
Safety and Handling
RTECSNumber : SL0590000
TSCA : No