missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Trimethylsilyl-1,3-dithiane 96.0+%, TCI America™
$344.86 - $1707.48
Chemical Identifiers
CAS | 13411-42-2 |
---|---|
Molecular Formula | C7H16S2Si |
Molecular Weight (g/mol) | 192.41 |
MDL Number | MFCD00006655 |
InChI Key | BTTUMVHWIAXYPJ-UHFFFAOYSA-N |
Synonym | 2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane |
PubChem CID | 83413 |
IUPAC Name | (1,3-dithian-2-yl)trimethylsilane |
SMILES | C[Si](C)(C)C1SCCCS1 |
Chemical Identifiers
13411-42-2 | |
192.41 | |
BTTUMVHWIAXYPJ-UHFFFAOYSA-N | |
83413 | |
C[Si](C)(C)C1SCCCS1 |
C7H16S2Si | |
MFCD00006655 | |
2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane | |
(1,3-dithian-2-yl)trimethylsilane |
Specifications
13411-42-2 | |
77°C | |
MFCD00006655 | |
2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane | |
C[Si](C)(C)C1SCCCS1 | |
192.41 | |
192.41 | |
Liquid |
Yellow | |
C7H16S2Si | |
5 g | |
BTTUMVHWIAXYPJ-UHFFFAOYSA-N | |
(1,3-dithian-2-yl)trimethylsilane | |
83413 | |
≥96.0% (GC) | |
2-Trimethylsilyl-1,3-dithiane |
Safety and Handling
TSCA : No