Learn More
2-Thiazolecarboxaldehyde, 98%
CAS: 10200-59-6 | C4H3NOS | 113.14 g/mol
$142.60 - $142.60
Chemical Identifiers
CAS | 10200-59-6 |
---|---|
InChI Key | ZGTFNNUASMWGTM-UHFFFAOYSA-N |
Synonym | 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929 |
PubChem CID | 2734903 |
ChEBI | CHEBI:43623 |
IUPAC Name | 1,3-thiazole-2-carbaldehyde |
SMILES | C1=CSC(=N1)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC377680010
|
Thermo Scientific Chemicals
377680010 |
1 g | Glass bottle |
Each for $142.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10200-59-6 | |
2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929 | |
CHEBI:43623 | |
C1=CSC(=N1)C=O |
ZGTFNNUASMWGTM-UHFFFAOYSA-N | |
2734903 | |
1,3-thiazole-2-carbaldehyde |
Specifications
10200-59-6 | |
1.2880g/mL | |
68°C | |
97.5% min. (GC) | |
C4H3NOS | |
1 g | |
2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929 | |
ZGTFNNUASMWGTM-UHFFFAOYSA-N | |
1,3-thiazole-2-carbaldehyde | |
2734903 | |
113.14 | |
Liquid |
Yellow to Brown | |
76.0°C to 78.0°C (24.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5735 to 1.5755 | |
1.288 | |
Solubility in water: soluble. | |
C1=CSC(=N1)C=O | |
113.14 | |
CHEBI:43623 | |
98% | |
2-Thiazolecarboxaldehyde |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
RUO – Research Use Only