missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$6340.67 - $6340.67
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Description
- For Laboratory,Research,or Manufacturing Use
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Specifications
67-63-0 | |
12.00°C | |
1L = 0.79kg | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
Meets ACS reagent chemical requirements |
1 L | |
Aluminum Bottle | |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1 | |
2-Propanol |