missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Phenylthio)thiophene, 97+%
CAS: 16718-12-0 | C10H8S2 | 192.29 g/mol
$130.11 - $508.26
Chemical Identifiers
CAS | 16718-12-0 |
---|---|
Molecular Formula | C10H8S2 |
Molecular Weight (g/mol) | 192.29 |
MDL Number | MFCD00052141 |
InChI Key | JQTBWKNYWACCRU-UHFFFAOYSA-N |
PubChem CID | 2778915 |
IUPAC Name | 2-phenylsulfanylthiophene |
SMILES | S(C1=CC=CS1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1370803
|
Thermo Scientific Chemicals
A1370803 |
1 g |
Each for $130.11
|
|
|||||
AAA1370806
|
Thermo Scientific Chemicals
A1370806 |
5 g |
Each for $508.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16718-12-0 | |
192.29 | |
JQTBWKNYWACCRU-UHFFFAOYSA-N | |
2-phenylsulfanylthiophene |
C10H8S2 | |
MFCD00052141 | |
2778915 | |
S(C1=CC=CS1)C1=CC=CC=C1 |
Specifications
16718-12-0 | |
C10H8S2 | |
1 g | |
JQTBWKNYWACCRU-UHFFFAOYSA-N | |
2-phenylsulfanylthiophene | |
2778915 | |
≥97% |
109°C to 110°C (0.4 mmHg) | |
MFCD00052141 | |
2-phenylthio thiophene, 2-phenylsulfanyl thiophene, 2-phenylthiothiophene, phenylthiophenyl sulfide, acmc-1bvxh, 2-thienyl phenylsulfide, thiophene,2-phenylthio, thiophene, 2-phenylthio | |
S(C1=CC=CS1)C1=CC=CC=C1 | |
192.29 | |
192.3 | |
2-(Phenylthio)thiophene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only