Learn More
2-Phenoxybenzaldehyde, 98%, Thermo Scientific™
$298.34 - $298.34
Chemical Identifiers
CAS | 19434-34-5 |
---|---|
Molecular Formula | C13H10O2 |
Molecular Weight (g/mol) | 198.22 |
MDL Number | MFCD00800666 |
InChI Key | IMPIIVKYTNMBCD-UHFFFAOYSA-N |
Synonym | o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384 |
PubChem CID | 88060 |
IUPAC Name | 2-phenoxybenzaldehyde |
SMILES | O=CC1=CC=CC=C1OC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5044803
|
Thermo Scientific Chemicals
H5044803 |
1 g |
Each for $298.34
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-Phenoxybenzaldehyde is used as a reagent in the synthesis of chiral 1,3,4-oxadiazol-2-ones as potent and highly selective FAAH inhibitors. Also used as a reagent in the synthesis of [4-(phenoxy)pyridin-3-yl]methylamines; a new class of selective noradrenaline reuptake inhibitors (NRI).
Solubility
Very slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
Chemical Identifiers
19434-34-5 | |
198.22 | |
IMPIIVKYTNMBCD-UHFFFAOYSA-N | |
88060 | |
O=CC1=CC=CC=C1OC1=CC=CC=C1 |
C13H10O2 | |
MFCD00800666 | |
o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384 | |
2-phenoxybenzaldehyde |
Specifications
19434-34-5 | |
MFCD00800666 | |
Air Sensitive | |
Very slightly soluble in water. | |
O=CC1=CC=CC=C1OC1=CC=CC=C1 | |
198.22 | |
198.22 | |
2-Phenoxybenzaldehyde |
C13H10O2 | |
1 g | |
o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384 | |
IMPIIVKYTNMBCD-UHFFFAOYSA-N | |
2-phenoxybenzaldehyde | |
88060 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 243-061-0
TSCA : No
Recommended Storage : Ambient temperatures