missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrocinnamaldehyde 98.0+%, TCI America™

$139.81 - $417.56

Chemical Identifiers

CAS 1466-88-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00007188
InChI Key VMSMELHEXDVEDE-HWKANZROSA-N
Synonym 2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
PubChem CID 5367122
IUPAC Name (E)-3-(2-nitrophenyl)prop-2-enal
SMILES C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06115G
View Documents
TCI America
N06115G
5 g
Each for $139.81
Only null left
 
N061125G
View Documents
TCI America
N061125G
25 g
Each for $417.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1466-88-2
177.159
VMSMELHEXDVEDE-HWKANZROSA-N
5367122
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
C9H7NO3
MFCD00007188
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
(E)-3-(2-nitrophenyl)prop-2-enal

Specifications

1466-88-2
C9H7NO3
5 g
VMSMELHEXDVEDE-HWKANZROSA-N
(E)-3-(2-nitrophenyl)prop-2-enal
5367122
≥98.0% (GC)
2-Nitrocinnamaldehyde
128°C
MFCD00007188
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
177.159
177.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'