missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)benzeneboronic acid, 98%

CAS: 330804-03-0 | C7H9BO4S | 200.015 g/mol

$236.55 - $917.96

Chemical Identifiers

CAS 330804-03-0
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.015
MDL Number MFCD02179472
InChI Key MSQFFCRGQPVQRS-UHFFFAOYSA-N
Synonym 2-methylsulfonyl phenylboronic acid, 2-methylsulfonyl phenyl boronic acid, 2-methylsulfonylphenyl boronic acid, 2-methanesulfonyl phenylboronic acid, 2-methylsulphonyl phenylboronic acid, 2-methanesulfonylphenylboronic acid, 2-methylsulphonyl benzeneboronic acid, 2-methylsulfonyl benzeneboronic acid, b-2-methylsulfonyl phenyl-boronic acid
PubChem CID 2773539
IUPAC Name (2-methylsulfonylphenyl)boronic acid
SMILES B(C1=CC=CC=C1S(=O)(=O)C)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5244503
View Documents
Thermo Scientific Chemicals
H5244503
1 g
Each for $236.55
Only null left
 
AAH5244506
View Documents
Thermo Scientific Chemicals
H5244506
5 g
Each for $917.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

330804-03-0
200.015
MSQFFCRGQPVQRS-UHFFFAOYSA-N
2773539
B(C1=CC=CC=C1S(=O)(=O)C)(O)O
C7H9BO4S
MFCD02179472
2-methylsulfonyl phenylboronic acid, 2-methylsulfonyl phenyl boronic acid, 2-methylsulfonylphenyl boronic acid, 2-methanesulfonyl phenylboronic acid, 2-methylsulphonyl phenylboronic acid, 2-methanesulfonylphenylboronic acid, 2-methylsulphonyl benzeneboronic acid, 2-methylsulfonyl benzeneboronic acid, b-2-methylsulfonyl phenyl-boronic acid
(2-methylsulfonylphenyl)boronic acid

Specifications

330804-03-0
C7H9BO4S
1 g
MSQFFCRGQPVQRS-UHFFFAOYSA-N
(2-methylsulfonylphenyl)boronic acid
2773539
98%
78°C to 84°C
MFCD02179472
2-methylsulfonyl phenylboronic acid, 2-methylsulfonyl phenyl boronic acid, 2-methylsulfonylphenyl boronic acid, 2-methanesulfonyl phenylboronic acid, 2-methylsulphonyl phenylboronic acid, 2-methanesulfonylphenylboronic acid, 2-methylsulphonyl benzeneboronic acid, 2-methylsulfonyl benzeneboronic acid, b-2-methylsulfonyl phenyl-boronic acid
B(C1=CC=CC=C1S(=O)(=O)C)(O)O
200.015
200.02
2-(Methylsulfonyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only