missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane 97.0+%, TCI America™

$42.87 - $68.97

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M016725ML
View Documents
TCI America
M016725ML
25 mL
Each for $42.87
Only null left
 
M0167500ML
View Documents
TCI America
M0167500ML
500 mL
Each for $68.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

78-78-4
72.15
QWTDNUCVQCZILF-UHFFFAOYSA-N
6556
2-methylbutane
C5H12
MFCD00009338
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
CHEBI:30362
CCC(C)C

Specifications

78-78-4
31°C
Clear Liquid at 20°C
MFCD00009338
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
CCC(C)C
72.15
CHEBI:30362
≥98.0% (GC)
Colorless
25 mL
C5H12
1265
QWTDNUCVQCZILF-UHFFFAOYSA-N
2-methylbutane
6556
72.15
2-Methylbutane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0005

RTECSNumber : EK4430000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'