missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptopyridine N-Oxide Sodium Salt Anhydrous 98.0+%, TCI America™
$166.30 - $570.71
Chemical Identifiers
CAS | 3811-73-2 |
---|---|
Molecular Formula | C5H4NNaOS |
Molecular Weight (g/mol) | 149.143 |
MDL Number | MFCD01941547 |
InChI Key | XNRNJIIJLOFJEK-UHFFFAOYSA-N |
Synonym | sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van |
PubChem CID | 19658 |
IUPAC Name | sodium;1-oxidopyridine-2-thione |
SMILES | C1=CC(=S)N(C=C1)[O-].[Na+] |
Chemical Identifiers
3811-73-2 | |
149.143 | |
XNRNJIIJLOFJEK-UHFFFAOYSA-N | |
19658 | |
C1=CC(=S)N(C=C1)[O-].[Na+] |
C5H4NNaOS | |
MFCD01941547 | |
sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van | |
sodium;1-oxidopyridine-2-thione |
Specifications
3811-73-2 | |
White-Yellow | |
MFCD01941547 | |
sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van | |
C1=CC(=S)N(C=C1)[O-].[Na+] | |
149.143 | |
149.14 | |
Crystalline Powder |
287°C | |
C5H4NNaOS | |
5 g | |
XNRNJIIJLOFJEK-UHFFFAOYSA-N | |
sodium;1-oxidopyridine-2-thione | |
19658 | |
≥98.0% (HPLC) | |
2-Mercaptopyridine N-Oxide Sodium Salt Anhydrous |
Safety and Handling
EINECSNumber : (9)-1473
RTECSNumber : UT9000000
TSCA : Yes