missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptoethanol 98.0+%, TCI America™
$35.90 - $83.14
Chemical Identifiers
CAS | 60-24-2 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00004890 |
InChI Key | DGVVWUTYPXICAM-UHFFFAOYSA-N |
Synonym | 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan |
PubChem CID | 1567 |
ChEBI | CHEBI:41218 |
IUPAC Name | 2-sulfanylethan-1-ol |
SMILES | OCCS |
Chemical Identifiers
60-24-2 | |
78.13 | |
DGVVWUTYPXICAM-UHFFFAOYSA-N | |
1567 | |
2-sulfanylethan-1-ol |
C2H6OS | |
MFCD00004890 | |
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
CHEBI:41218 | |
OCCS |
Specifications
60-24-2 | |
Colorless | |
C2H6OS | |
25 g | |
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
OCCS | |
78.13 | |
CHEBI:41218 | |
≥98.0% (GC) | |
2-Mercaptoethanol |
-100°C | |
158°C | |
MFCD00004890 | |
2966 | |
DGVVWUTYPXICAM-UHFFFAOYSA-N | |
2-sulfanylethan-1-ol | |
1567 | |
78.13 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0458
RTECSNumber : KL5600000
TSCA : Yes