missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol, Spectrum™ Chemical
$316.30 - $15202.10
Chemical Identifiers
CAS | 110-80-5 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
InChI Key | ZNQVEEAIQZEUHB-UHFFFAOYSA-N |
IUPAC Name | 2-ethoxyethan-1-ol |
SMILES | CCOCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18608207
|
Spectrum Chemical Mfg Cor
E10211LT |
1 L | Amber Glass Bottle |
Each for $316.30
|
|
||||
18608210
|
Spectrum Chemical Mfg Cor
E10214LTGL |
4 L | Amber Glass Bottle |
Each for $723.50
|
|
||||
18608208
|
Spectrum Chemical Mfg Cor
E102120LTMT |
20 L | Steel Can |
Each for $2,551.17
|
|
||||
18608209
|
Spectrum Chemical Mfg Cor
E1021200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
110-80-5 | |
90.12 | |
2-ethoxyethan-1-ol |
C4H10O2 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
CCOCCO |
Specifications
5e-05 | |
1 | |
C4H10O2 | |
0.9290 - 0.9330 | |
CCOCCO | |
90.12 |
110-80-5 | |
Amber Glass Bottle | |
1 L | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
2-ethoxyethan-1-ol | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.