missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenothiazine 98.0+%, TCI America™
$59.65 - $576.00
Chemical Identifiers
CAS | 92-39-7 |
---|---|
Molecular Formula | C12H8ClNS |
Molecular Weight (g/mol) | 233.713 |
MDL Number | MFCD00005016 |
InChI Key | KFZGLJSYQXZIGP-UHFFFAOYSA-N |
Synonym | 2-chlorophenothiazine, 10h-phenothiazine, 2-chloro, phenothiazine, 2-chloro, 2-chlorothiodiphenylamine, wln: t c666 bm isj eg, 2-chloro-phenothiazine, 2-chloropheno-thiazine, acmc-209rfl, 2-chlorophenothizine, dsstox_cid_31643 |
PubChem CID | 7088 |
IUPAC Name | 2-chloro-10H-phenothiazine |
SMILES | C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)Cl |
Chemical Identifiers
92-39-7 | |
233.713 | |
KFZGLJSYQXZIGP-UHFFFAOYSA-N | |
7088 | |
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)Cl |
C12H8ClNS | |
MFCD00005016 | |
2-chlorophenothiazine, 10h-phenothiazine, 2-chloro, phenothiazine, 2-chloro, 2-chlorothiodiphenylamine, wln: t c666 bm isj eg, 2-chloro-phenothiazine, 2-chloropheno-thiazine, acmc-209rfl, 2-chlorophenothizine, dsstox_cid_31643 | |
2-chloro-10H-phenothiazine |
Specifications
92-39-7 | |
C12H8ClNS | |
25 g | |
KFZGLJSYQXZIGP-UHFFFAOYSA-N | |
2-chloro-10H-phenothiazine | |
7088 | |
≥98.0% (GC) | |
2-Chlorophenothiazine |
198°C | |
MFCD00005016 | |
2-chlorophenothiazine, 10h-phenothiazine, 2-chloro, phenothiazine, 2-chloro, 2-chlorothiodiphenylamine, wln: t c666 bm isj eg, 2-chloro-phenothiazine, 2-chloropheno-thiazine, acmc-209rfl, 2-chlorophenothizine, dsstox_cid_31643 | |
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)Cl | |
233.713 | |
233.71 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3803
RTECSNumber : SN7580000
TSCA : Yes