missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzylideneacetone, 98%

CAS: 175136-82-0 | C10H8ClFO | 198.621 g/mol

$49.07 - $118.99

Chemical Identifiers

CAS 175136-82-0
Molecular Formula C10H8ClFO
Molecular Weight (g/mol) 198.621
MDL Number MFCD00051608
InChI Key VZRBDBXFAKDNDJ-AATRIKPKSA-N
Synonym 2-chloro-6-fluorobenzylideneacetone, 3e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one, e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one, labotest-bb lt00455271, 1-2-chloro-6-fluorophenyl-1-buten-2-one, e-4-2-chloro-6-fluorophenyl-3-buten-2-one, e-4-2-chloro-6-fluorophenyl but-3-en-2-one
PubChem CID 5373895
IUPAC Name (E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=C(C=CC=C1Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1508303
View Documents
Thermo Scientific Chemicals
A1508303
1 g
Each for $49.07
Only null left
 
AAA1508306
View Documents
Thermo Scientific Chemicals
A1508306
5 g
Each for $118.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

175136-82-0
198.621
VZRBDBXFAKDNDJ-AATRIKPKSA-N
5373895
CC(=O)C=CC1=C(C=CC=C1Cl)F
C10H8ClFO
MFCD00051608
2-chloro-6-fluorobenzylideneacetone, 3e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one, e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one, labotest-bb lt00455271, 1-2-chloro-6-fluorophenyl-1-buten-2-one, e-4-2-chloro-6-fluorophenyl-3-buten-2-one, e-4-2-chloro-6-fluorophenyl but-3-en-2-one
(E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one

Specifications

175136-82-0
C10H8ClFO
MFCD00051608
2-chloro-6-fluorobenzylideneacetone, 3e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one, e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one, labotest-bb lt00455271, 1-2-chloro-6-fluorophenyl-1-buten-2-one, e-4-2-chloro-6-fluorophenyl-3-buten-2-one, e-4-2-chloro-6-fluorophenyl but-3-en-2-one
CC(=O)C=CC1=C(C=CC=C1Cl)F
198.621
198.63
2-Chloro-6-fluorobenzylideneacetone
115°C (0.8 mmHg)
1.58
1 g
VZRBDBXFAKDNDJ-AATRIKPKSA-N
(E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
5373895
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only