missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-2-oxo-1,3,2-dioxaphospholane 95.0+%, TCI America™
$158.44 - $486.52
Chemical Identifiers
CAS | 6609-64-9 |
---|---|
Molecular Formula | C2H4ClO3P |
Molecular Weight (g/mol) | 142.475 |
MDL Number | MFCD00043138 |
InChI Key | SBMUNILHNJLMBF-UHFFFAOYSA-N |
Synonym | 2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate |
PubChem CID | 81087 |
IUPAC Name | 2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide |
SMILES | C1COP(=O)(O1)Cl |
Chemical Identifiers
6609-64-9 | |
142.475 | |
SBMUNILHNJLMBF-UHFFFAOYSA-N | |
81087 | |
C1COP(=O)(O1)Cl |
C2H4ClO3P | |
MFCD00043138 | |
2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate | |
2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide |
Specifications
6609-64-9 | |
91°C | |
MFCD00043138 | |
3265 | |
SBMUNILHNJLMBF-UHFFFAOYSA-N | |
2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide | |
81087 | |
≥95.0% (GC,T) | |
2-Chloro-2-oxo-1,3,2-dioxaphospholane |
Yellow | |
C2H4ClO3P | |
5 g | |
2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate | |
C1COP(=O)(O1)Cl | |
142.475 | |
142.47 | |
Liquid |
Safety and Handling
EINECSNumber : (9)-0448
TSCA : No
Recommended Storage : Freezer