missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)
$182.08 - $851.48
Specifications
Melting Point | -87°C |
---|---|
Density | 0.805g/mL at 25°C |
Boiling Point | 79°C to 81°C |
Quantity | 1 L |
Flash Point | −3°C (26.6°F) |
Odor | Solvent like |
Packaging | Glass Bottle |
Solubility Information | 290g/L at 20°C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
60046232
|
Honeywell Inc
334071L |
1 L | Glass Bottle |
Each for $182.08
|
|
||||
60046233
|
Honeywell Inc
3340725L |
2.5 L | Glass Bottle |
Each for $276.83
|
|
||||
60046236
|
Honeywell Inc
334074X25L |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $851.48
|
|
||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
-87°C | |
79°C to 81°C | |
−3°C (26.6°F) | |
Glass Bottle |
0.805g/mL at 25°C | |
1 L | |
Solvent like | |
290g/L at 20°C |