missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,4-dichlorobenzene, 98%

CAS: 1435-50-3 | C6H3BrCl2 | 225.894 g/mol

$76.23 - $241.60

Chemical Identifiers

CAS 1435-50-3
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00018505
InChI Key OVXVQBCRONSPDC-UHFFFAOYSA-N
Synonym 1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
PubChem CID 15033
IUPAC Name 2-bromo-1,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1205606
View Documents
Thermo Scientific Chemicals
A1205606
5 g
Each for $76.23
Only null left
 
AAA1205614
View Documents
Thermo Scientific Chemicals
A1205614
25 g
Each for $241.60
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1435-50-3
225.894
OVXVQBCRONSPDC-UHFFFAOYSA-N
15033
C1=CC(=C(C=C1Cl)Br)Cl
C6H3BrCl2
MFCD00018505
1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
2-bromo-1,4-dichlorobenzene

Specifications

1435-50-3
118°C to 120°C (20 mmHg)
C6H3BrCl2
5 g
1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
C1=CC(=C(C=C1Cl)Br)Cl
225.894
225.91
2-Bromo-1,4-dichlorobenzene
32°C to 36°C
>110°C (230°F)
MFCD00018505
1934815
OVXVQBCRONSPDC-UHFFFAOYSA-N
2-bromo-1,4-dichlorobenzene
15033
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 215-859-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only