Learn More
2-Aminothiobenzamide, 97%
CAS: 2454-39-9 | C7H8N2S | 152.215 g/mol
$85.62 - $285.99
Chemical Identifiers
CAS | 2454-39-9 |
---|---|
Molecular Formula | C7H8N2S |
Molecular Weight (g/mol) | 152.215 |
MDL Number | MFCD00963496 |
InChI Key | HPZKAJRFABCGFF-UHFFFAOYSA-N |
Synonym | 2-aminothiobenzamide, 2-amino-thiobenzamide, 2-aminobenzothioamide, benzenecarbothioamide, 2-amino, thioanthranilamide, o-aminothiobenzamide, 2-aminobenzene-1-carbothioamide, amino 2-aminophenyl methane-1-thione, benzamide, o-aminothio, thioanthranilsaureamid |
PubChem CID | 1550990 |
IUPAC Name | 2-aminobenzenecarbothioamide |
SMILES | C1=CC=C(C(=C1)C(=S)N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5180903
|
Thermo Scientific Chemicals
H5180903 |
1 g |
Each for $85.62
|
|
|||||
AAH5180906
|
Thermo Scientific Chemicals
H5180906 |
5 g |
Each for $285.99
|
|
|||||
Description
It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
2454-39-9 | |
152.215 | |
HPZKAJRFABCGFF-UHFFFAOYSA-N | |
1550990 | |
C1=CC=C(C(=C1)C(=S)N)N |
C7H8N2S | |
MFCD00963496 | |
2-aminothiobenzamide, 2-amino-thiobenzamide, 2-aminobenzothioamide, benzenecarbothioamide, 2-amino, thioanthranilamide, o-aminothiobenzamide, 2-aminobenzene-1-carbothioamide, amino 2-aminophenyl methane-1-thione, benzamide, o-aminothio, thioanthranilsaureamid | |
2-aminobenzenecarbothioamide |
Specifications
2454-39-9 | |
C7H8N2S | |
1 g | |
Slightly soluble in water. | |
C1=CC=C(C(=C1)C(=S)N)N | |
152.215 | |
152.21 | |
2-Aminothiobenzamide |
115°C to 120°C | |
MFCD00963496 | |
2-aminothiobenzamide, 2-amino-thiobenzamide, 2-aminobenzothioamide, benzenecarbothioamide, 2-amino, thioanthranilamide, o-aminothiobenzamide, 2-aminobenzene-1-carbothioamide, amino 2-aminophenyl methane-1-thione, benzamide, o-aminothio, thioanthranilsaureamid | |
HPZKAJRFABCGFF-UHFFFAOYSA-N | |
2-aminobenzenecarbothioamide | |
1550990 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only