missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanol, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor ET1001LTGL

 View more versions of this product

Catalog No. 18608192


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

141-43-5
Amber Glass Bottle
1 L
NCCO
61.08
ACS
1
C2H7NO
HZAXFHJVJLSVMW-UHFFFAOYSA-N
2-aminoethan-1-ol
0.99

Chemical Identifiers

141-43-5
61.08
2-aminoethan-1-ol
C2H7NO
HZAXFHJVJLSVMW-UHFFFAOYSA-N
NCCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.