missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-nitropyridine 98.0+%, TCI America™
$128.62 - $777.32
Chemical Identifiers
CAS | 4214-76-0 |
---|---|
Molecular Formula | C5H5N3O2 |
Molecular Weight (g/mol) | 139.11 |
MDL Number | MFCD00006325 |
InChI Key | UGSBCCAHDVCHGI-UHFFFAOYSA-N |
Synonym | 2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin |
PubChem CID | 77888 |
IUPAC Name | 5-nitropyridin-2-amine |
SMILES | NC1=CC=C(C=N1)[N+]([O-])=O |
Chemical Identifiers
4214-76-0 | |
139.11 | |
UGSBCCAHDVCHGI-UHFFFAOYSA-N | |
77888 | |
NC1=CC=C(C=N1)[N+]([O-])=O |
C5H5N3O2 | |
MFCD00006325 | |
2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin | |
5-nitropyridin-2-amine |
Specifications
4214-76-0 | |
Yellow | |
MFCD00006325 | |
2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin | |
NC1=CC=C(C=N1)[N+]([O-])=O | |
139.11 | |
139.11 | |
Crystalline Powder |
188°C | |
C5H5N3O2 | |
25 g | |
UGSBCCAHDVCHGI-UHFFFAOYSA-N | |
5-nitropyridin-2-amine | |
77888 | |
≥98.0% (T) | |
2-Amino-5-nitropyridine |
Safety and Handling
TSCA : Yes