missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,9-Diacetylguanine, 96%, Thermo Scientific™
$53.93 - $53.93
Chemical Identifiers
CAS | 3056-33-5 |
---|---|
Molecular Formula | C9H9N5O3 |
Molecular Weight (g/mol) | 235.20 |
MDL Number | MFCD00142116 |
InChI Key | GILZZWCROUGLIS-UHFFFAOYSA-N |
PubChem CID | 76461 |
IUPAC Name | N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide |
SMILES | CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC459210250
|
Thermo Scientific Chemicals
459210250 |
25 g |
Each for $53.93
|
|
|||||
Chemical Identifiers
3056-33-5 | |
235.20 | |
GILZZWCROUGLIS-UHFFFAOYSA-N | |
N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide |
C9H9N5O3 | |
MFCD00142116 | |
76461 | |
CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O |
Specifications
3056-33-5 | |
Authentic | |
MFCD00142116 | |
n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide | |
CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O | |
235.20 | |
235.2 | |
2,9-Diacetylguanine |
285.0°C | |
C9H9N5O3 | |
25 g | |
GILZZWCROUGLIS-UHFFFAOYSA-N | |
N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide | |
76461 | |
96% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 221-287-
RUO – Research Use Only