Learn More
2,7-Dibromocarbazole
CAS: 136630-39-2 | C12H7Br2N | 325.00 g/mol
$118.19 - $474.64
Chemical Identifiers
CAS | 136630-39-2 |
---|---|
Molecular Formula | C12H7Br2N |
Molecular Weight (g/mol) | 325.00 |
InChI Key | QPTWWBLGJZWRAV-UHFFFAOYSA-N |
IUPAC Name | 2,7-dibromo-9H-carbazole |
SMILES | BrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC467780010
|
Thermo Scientific Chemicals
467780010 |
1 g |
Each for $118.19
|
|
|||||
AC467780050
|
Thermo Scientific Chemicals
467780050 |
5 g |
Each for $474.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
136630-39-2 | |
325.00 | |
2,7-dibromo-9H-carbazole |
C12H7Br2N | |
QPTWWBLGJZWRAV-UHFFFAOYSA-N | |
BrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1 |
Specifications
136630-39-2 | |
C12H7Br2N | |
QPTWWBLGJZWRAV-UHFFFAOYSA-N | |
2,7-dibromo-9H-carbazole | |
325 |
225.0°C to 230.0°C | |
1 g | |
BrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1 | |
325.00 | |
2,7-Dibromocarbazole |
Safety and Handling
GHS H Statement Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
RUO – Research Use Only