missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic Dianhydride 98.0+%, TCI America™
$423.14 - $1446.51
Chemical Identifiers
CAS | 83204-68-6 |
---|---|
Molecular Formula | C14H2Br2O6 |
Molecular Weight (g/mol) | 425.972 |
MDL Number | MFCD00406909 |
InChI Key | FEDCHNPLDKDTNS-UHFFFAOYSA-N |
PubChem CID | 11974609 |
SMILES | C1=C2C3=C4C(=CC(=C3C(=O)OC2=O)Br)C(=O)OC(=O)C4=C1Br |
Chemical Identifiers
83204-68-6 | |
425.972 | |
FEDCHNPLDKDTNS-UHFFFAOYSA-N | |
C1=C2C3=C4C(=CC(=C3C(=O)OC2=O)Br)C(=O)OC(=O)C4=C1Br |
C14H2Br2O6 | |
MFCD00406909 | |
11974609 |
Specifications
83204-68-6 | |
Yellow | |
MFCD00406909 | |
FEDCHNPLDKDTNS-UHFFFAOYSA-N | |
425.972 | |
425.97 | |
Crystalline Powder |
350°C | |
C14H2Br2O6 | |
1 g | |
C1=C2C3=C4C(=CC(=C3C(=O)OC2=O)Br)C(=O)OC(=O)C4=C1Br | |
11974609 | |
≥98.0% (HPLC,T) | |
2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic Dianhydride |
Safety and Handling
TSCA : No