missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene 98.0+%, TCI America™

$199.46 - $689.70

Chemical Identifiers

CAS 67061-69-2
Molecular Formula C8H2Br2S3
Molecular Weight (g/mol) 354.092
InChI Key UTKPZZGBROTDKR-UHFFFAOYSA-N
PubChem CID 11382649
IUPAC Name 2,6-dibromodithieno[2,3-a
SMILES C1=C(SC2=C1SC3=C2SC(=C3)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D3799200MG
View Documents
TCI America
D3799200MG
200 mg
Each for $199.46
Only null left
 
D37991G
View Documents
TCI America
D37991G
1 g
Each for $689.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67061-69-2
354.092
11382649
C1=C(SC2=C1SC3=C2SC(=C3)Br)Br
C8H2Br2S3
UTKPZZGBROTDKR-UHFFFAOYSA-N
2,6-dibromodithieno[2,3-a

Specifications

67061-69-2
C8H2Br2S3
UTKPZZGBROTDKR-UHFFFAOYSA-N
2,6-dibromodithieno[2,3-a
11382649
≥98.0% (GC)
2,6-Dibromodithieno[3,2-b:2′,3′-d]thiophene
171°C
200 mg
C1=C(SC2=C1SC3=C2SC(=C3)Br)Br
354.092
354.09
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'